Gene Transcripts
- Gene Symbol
- FARP2
- Entrez Gene ID
- 9855
- Gene Name
- FERM, ARH/RhoGEF and pleckstrin domain protein 2
- Gene Aliases
- FIR, FRG, PLEKHC3
- Chromosome Location
- Chr. 2: 241356249 - 241494842 on Build GRCh38
- UniGene ID
- Hs.726316
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282983.1 |
NP_001269912.1
|
6, 7 |
682 |
|
NM_001282984.1 |
NP_001269913.1
|
6, 7 |
682 |
|
NM_014808.3 |
NP_055623.1
|
6, 7 |
682 |
|
XM_005247050.3 |
XP_005247107.1
|
6, 7 |
646 |
|
XM_011512233.1 |
XP_011510535.1
|
6, 7 |
646 |
|
XM_011512234.1 |
XP_011510536.1
|
6, 7 |
571 |
|
XM_011512235.1 |
XP_011510537.1
|
7, 8 |
904 |
|
XM_011512236.1 |
XP_011510538.1
|
6, 7 |
700 |
|
XM_017005416.1 |
XP_016860905.1
|
7, 8 |
861 |
|
XM_017005417.1 |
XP_016860906.1
|
7, 8 |
861 |
|
XM_017005418.1 |
XP_016860907.1
|
4, 5 |
499 |
|
XM_017005419.1 |
XP_016860908.1
|
6, 7 |
646 |
|
XM_017005420.1 |
XP_016860909.1
|
6, 7 |
646 |
|
GenBank mRNA |
AB018336.1 |
- |
6, 7 |
635 |
|
AK300435.1 |
- |
6, 7 |
642 |
|
BC021301.2 |
- |
6, 7 |
634 |
5013180 |
View Full Product Details