Gene Transcripts

Gene Symbol
NUAK1
Entrez Gene ID
9891
Gene Name
NUAK family kinase 1
Gene Aliases
ARK5
Chromosome Location
Chr. 12: 106063347 - 106140033 on Build GRCh38
UniGene ID
Hs.524692
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014840.2 NP_055655.1 7 2617
GenBank mRNA AB011109.1 - 7 2617
AK289992.1 - 7 1297
BC152462.1 - 7 2617 8862598

View Full Product Details