Gene Transcripts

Gene Symbol
ALG3
Entrez Gene ID
10195
Gene Name
ALG3, alpha-1,3- mannosyltransferase
Gene Aliases
CDG1D, CDGS4, CDGS6, D16Ertd36e, NOT56L, Not56, not
Chromosome Location
Chr. 3: 184242329 - 184249525 on Build GRCh38
UniGene ID
Hs.478481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006941.2 NP_001006942.1 2 353
NM_005787.5 NP_005778.1 2 280
NR_024534.1 2 243
XM_011512322.1 XP_011510624.1 2 165
XM_011512323.2 XP_011510625.1 2 151
GenBank mRNA AK289361.1 - 2 309
BC002839.2 - 2 255 3636604
BC004313.1 - 2 235
BC021062.1 - 2 244
BM920198.1 - 2 209
BX339649.2 - 2 358
KP189279.1 - 2 273
KP189280.1 - 2 236
KP189282.1 - 2 390
KP189284.1 - 2 271
KP189285.1 - 2 234
KP189287.1 - 2 189
KP189289.1 - 2 270
KP189290.1 - 2 234
KP189292.1 - 2 189
KP189294.1 - 2 249
KP189295.1 - 2 249
KP189296.1 - 2 212
KP189297.1 - 2 212
KP189299.1 - 2 105
KP189301.1 - 2 212
KP189302.1 - 2 270
KP189303.1 - 2 232
KP189305.1 - 2 189
KP189307.1 - 2 269
KP189308.1 - 2 232
KP189310.1 - 2 189
KP189312.1 - 2 269
KP189313.1 - 2 269
KP189314.1 - 2 269
KP189315.1 - 2 232
KP189316.1 - 2 232
KP189317.1 - 2 232
KP189321.1 - 2 189
KP189322.1 - 2 189
KP189323.1 - 2 189
Y09022.1 - 2 280

View Full Product Details