Gene Transcripts

Gene Symbol
ERLIN1
Entrez Gene ID
10613
Gene Name
ER lipid raft associated 1
Gene Aliases
C10orf69, Erlin-1, KE04, KEO4, SPFH1, SPG62
Chromosome Location
Chr. 10: 100150090 - 100186057 on Build GRCh38
UniGene ID
Hs.150087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100626.1 NP_001094096.1 6 445
NM_006459.3 NP_006450.2 5 689
XM_005269442.3 XP_005269499.1 6 475
XM_017015541.1 XP_016871030.1 6 459
GenBank mRNA AF064093.1 - 6 421
AK056574.1 - 5 3046
AK222480.1 - 6 417
AK298427.1 - 5 335
AK315580.1 - 6 417
BC031791.1 - 5 358 4214940
BP280488.1 - 6 445

View Full Product Details