Gene Transcripts

Gene Symbol
PHTF1
Entrez Gene ID
10745
Gene Name
putative homeodomain transcription factor 1
Gene Aliases
PHTF
Chromosome Location
Chr. 1: 113697202 - 113759888 on Build GRCh38
UniGene ID
Hs.655824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323041.1 NP_001309970.1 4 802
NM_001323042.1 NP_001309971.1 4 286
NM_001323043.1 NP_001309972.1 4 687
NM_001323044.1 NP_001309973.1 4 1060
NM_001323045.1 NP_001309974.1 5 771
NM_001323047.1 NP_001309976.1 4 802
NM_001323048.1 NP_001309977.1 4 687
NM_001323049.1 NP_001309978.1 3 976
NM_001323050.1 NP_001309979.1 5 771
NM_001323051.1 NP_001309980.1 4 1060
NM_001323052.1 NP_001309981.1 4 687
NM_001323053.1 NP_001309982.1 4 802
NM_006608.2 NP_006599.2 3 591
NR_136564.1 5 886
NR_136565.1 4 687
NR_136566.1 4 802
NR_136567.1 4 687
NR_136568.1 5 771
GenBank mRNA AJ011863.1 - 3 170
AK294749.1 - 3 317
BC002447.2 - 4 735 3346990
CX763183.1 - 3 467

View Full Product Details