Gene Transcripts

Gene Symbol
SF3A3
Entrez Gene ID
10946
Gene Name
splicing factor 3a subunit 3
Gene Aliases
PRP9, PRPF9, SAP61, SF3a60
Chromosome Location
Chr. 1: 37956975 - 37990110 on Build GRCh38
UniGene ID
Hs.77897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320830.1 NP_001307759.1 5 485
NM_006802.3 NP_006793.1 7 644
GenBank mRNA AK128438.1 - 7 1453
AK222554.1 - 7 520
AK291181.1 - 7 525
AK301424.1 - 5 362
BC002395.2 - 7 537 2821350
BC011523.1 - 7 601 3867442
U08815.1 - 7 507

View Full Product Details