Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014272.4 | NP_055087.2 | 3, 4 | 855 | |
XM_005254137.3 | XP_005254194.1 | 3, 4 | 856 | ||
GenBank mRNA | AB209497.1 | - | 2, 3 | 509 | |
AF140675.1 | - | 3, 4 | 624 | ||
AK299844.1 | - | 4, 5 | 790 | ||
AY327122.1 | - | 3, 4 | 622 | ||
BC061631.1 | - | 3, 4 | 835 | 6650221 |