Gene Transcripts

Gene Symbol
WDR6
Entrez Gene ID
11180
Gene Name
WD repeat domain 6
Gene Aliases
Chromosome Location
Chr. 3: 49007062 - 49015953 on Build GRCh38
UniGene ID
Hs.654815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320546.1 NP_001307475.1 2 995
NM_001320547.1 NP_001307476.1 2 366
NM_018031.4 NP_060501.3 2 781
GenBank mRNA AF099100.1 - 2 450
AK001080.1 - 2 450
AK225653.1 - 2 444
AK295145.1 - 2 781
AK304117.1 - 2 366
AK309913.1 - 2 661
BC101707.1 - 2 440 8069213
BC113467.1 - 2 440 8322519
BC143486.1 - 2 440
DC300417.1 - 2 367
DC389885.1 - 2 534

View Full Product Details