Gene Transcripts

Gene Symbol
RASSF1
Entrez Gene ID
11186
Gene Name
Ras association domain family member 1
Gene Aliases
123F2, NORE2A, RASSF1A, RDA32, REH3P21
Chromosome Location
Chr. 3: 50329786 - 50340936 on Build GRCh38
UniGene ID
Hs.476270
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206957.1 NP_001193886.1 5 996
NM_007182.4 NP_009113.3 6 1103
NM_170712.2 NP_733830.1 6 813
NM_170713.2 NP_733831.1 5 907
NM_170714.1 NP_733832.1 6 1115
XM_011533315.1 XP_011531617.1 7 1192
XM_011533316.2 XP_011531618.1 5 1011
GenBank mRNA AF040703.2 - 5 820
AF061836.1 - 5 903
AF102770.2 - 6 1008
AF102771.2 - 6 1020
AF102772.2 - 6 1020
AF132675.1 - 6 1010
AF132676.1 - 5 773
AF132677.1 - 6 800
AF286217.1 - 5 901
AF291719.1 - 4 796
AK292638.1 - 5 894
BC110412.1 - 6 813 5756246
BC117153.1 - 6 1079 40125704
BC143879.1 - 6 1079 9052395
BT020047.1 - 5 762
BT020048.1 - 5 762

View Full Product Details