Gene Transcripts
- Gene Symbol
- FNDC3A
- Entrez Gene ID
- 22862
- Gene Name
- fibronectin type III domain containing 3A
- Gene Aliases
- FNDC3, HUGO, bA203I16.1, bA203I16.5
- Chromosome Location
- Chr. 13: 48975241 - 49209779 on Build GRCh38
- UniGene ID
- Hs.508010
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001079673.1 |
NP_001073141.1
|
10, 11 |
1491 |
|
NM_001278438.1 |
NP_001265367.1
|
10, 11 |
1534 |
|
NM_014923.4 |
NP_055738.3
|
8, 9 |
1115 |
|
NR_103528.1 |
|
10, 11 |
1534 |
|
XM_011534997.2 |
XP_011533299.1
|
9, 10 |
1205 |
|
XM_017020440.1 |
XP_016875929.1
|
10, 11 |
1376 |
|
XM_017020441.1 |
XP_016875930.1
|
12, 13 |
1997 |
|
XM_017020442.1 |
XP_016875931.1
|
11, 12 |
1529 |
|
XM_017020443.1 |
XP_016875932.1
|
10, 11 |
1849 |
|
GenBank mRNA |
AB023187.2 |
- |
8, 9 |
1045 |
|
AJ749706.1 |
- |
10, 11 |
1493 |
|
AJ749707.1 |
- |
8, 9 |
1106 |
|
AK302415.1 |
- |
8, 9 |
1115 |
|
BC060816.1 |
- |
1, 2 |
153 |
|
BC070072.1 |
- |
1, 2 |
153 |
|
BC132812.1 |
- |
10, 11 |
1258 |
40146834 |
BC136617.1 |
- |
10, 11 |
1259 |
9020607 |
BC144301.1 |
- |
10, 11 |
1259 |
9052821 |
BX640739.1 |
- |
10, 11 |
1394 |
|
BX648141.1 |
- |
10, 11 |
1481 |
|
View Full Product Details