Gene Transcripts

Gene Symbol
LINC01565
Entrez Gene ID
23434
Gene Name
long intergenic non-protein coding RNA 1565
Gene Aliases
C3orf27, GR6
Chromosome Location
Chr. 3: 128572000 - 128576086 on Build GRCh38
UniGene ID
Hs.194283
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_125802.1 3 980
GenBank mRNA AF008192.1 - 3 980

View Full Product Details