Gene Transcripts

Gene Symbol
SLC35A3
Entrez Gene ID
23443
Gene Name
solute carrier family 35 member A3
Gene Aliases
AMRS
Chromosome Location
Chr. 1: 99969367 - 100026979 on Build GRCh38
UniGene ID
Hs.448979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271685.1 NP_001258614.1 6 951
NM_012243.2 NP_036375.1 6 1116
XM_005270691.4 XP_005270748.1 7 1637
XM_011541135.2 XP_011539437.1 5 970
XM_011541136.2 XP_011539438.1 7 1033
XM_011541137.2 XP_011539439.1 6 1093
XM_017000869.1 XP_016856358.1 5 1415
XM_017000870.1 XP_016856359.1 6 1538
XM_017000871.1 XP_016856360.1 5 1415
XM_017000872.1 XP_016856361.1 5 1383
GenBank mRNA AB021981.1 - 6 921
AK024989.1 - 5 682
AK290573.1 - 6 874
CR749816.1 - 6 951

View Full Product Details