Gene Transcripts
- Gene Symbol
- DNPEP
- Entrez Gene ID
- 23549
- Gene Name
- aspartyl aminopeptidase
- Gene Aliases
- ASPEP, DAP
- Chromosome Location
- Chr. 2: 219372029 - 219400022 on Build GRCh38
- UniGene ID
- Hs.258551
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319116.1 |
NP_001306045.1
|
3, 4 |
339 |
|
NM_001319117.1 |
NP_001306046.1
|
3, 4 |
339 |
|
NM_001319118.1 |
NP_001306047.1
|
3, 4 |
267 |
|
NM_001319119.1 |
NP_001306048.1
|
3, 4 |
547 |
|
NM_001319120.1 |
NP_001306049.1
|
3, 4 |
364 |
|
NM_001319121.1 |
NP_001306050.1
|
4, 5 |
240 |
|
NM_001319122.1 |
NP_001306051.1
|
3, 4 |
332 |
|
NM_012100.3 |
NP_036232.2
|
3, 4 |
364 |
|
NR_134970.1 |
|
3, 4 |
364 |
|
GenBank mRNA |
AF005050.2 |
- |
3, 4 |
339 |
|
AK001777.1 |
- |
3, 4 |
364 |
|
AK001977.1 |
- |
3, 4 |
336 |
|
AK021871.1 |
- |
3, 4 |
216 |
|
AK023005.1 |
- |
3, 4 |
278 |
|
AK301919.1 |
- |
3, 4 |
324 |
|
AK302809.1 |
- |
3, 4 |
331 |
|
BC000653.2 |
- |
3, 4 |
246 |
3343505 |
BI256589.1 |
- |
3, 4 |
230 |
|
BI821651.1 |
- |
3, 4 |
252 |
|
BI830094.1 |
- |
3, 4 |
343 |
|
BM564135.1 |
- |
3, 4 |
327 |
|
CN356162.1 |
- |
3, 4 |
258 |
|
DA203030.1 |
- |
3, 4 |
364 |
|
DA398660.1 |
- |
3, 4 |
267 |
|
View Full Product Details