Gene Transcripts
- Gene Symbol
- PAMR1
- Entrez Gene ID
- 25891
- Gene Name
- peptidase domain containing associated with muscle regeneration 1
- Gene Aliases
- DKFZP586H2123, FP938, RAMP
- Chromosome Location
- Chr. 11: 35431827 - 35530300 on Build GRCh38
- UniGene ID
- Hs.55044
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001001991.2 |
NP_001001991.1
|
8, 9 |
1545 |
|
NM_001282675.1 |
NP_001269604.1
|
10, 11 |
1535 |
|
NM_001282676.1 |
NP_001269605.1
|
6, 7 |
1212 |
|
NM_015430.3 |
NP_056245.2
|
9, 10 |
1596 |
|
GenBank mRNA |
AF370388.1 |
- |
7, 8 |
1034 |
|
AK027841.1 |
- |
9, 10 |
1196 |
|
AK297092.1 |
- |
10, 11 |
1536 |
|
AK300185.1 |
- |
6, 7 |
797 |
|
AK316188.1 |
- |
6, 7 |
784 |
|
AL832391.1 |
- |
8, 9 |
1111 |
|
AY358346.1 |
- |
8, 9 |
1124 |
|
BC089434.1 |
- |
9, 10 |
1188 |
30558617 |
BX640676.1 |
- |
8, 9 |
1545 |
|
View Full Product Details