Gene Transcripts

Gene Symbol
MTG2
Entrez Gene ID
26164
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
GTPBP5, ObgH1, dJ1005F21.2
Chromosome Location
Chr. 20: 62183025 - 62203568 on Build GRCh38
UniGene ID
Hs.340636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015666.3 NP_056481.1 7 1065
XM_005260393.1 XP_005260450.1 7 1055
XM_006723774.3 XP_006723837.1 8 1149
XM_011528776.1 XP_011527078.1 7 3069
XM_017027805.1 XP_016883294.1 6 3219
XM_017027806.1 XP_016883295.1 7 1054
XM_017027807.1 XP_016883296.1 6 3245
XM_017027808.1 XP_016883297.1 7 1081
GenBank mRNA AK001603.1 - 6 1045
AK124572.1 - 7 3356
AK299147.1 - 5 723
AK299726.1 - 4 574
AK316027.1 - 6 845
AK316325.1 - 7 917
AL117498.1 - 1 94
BC012073.1 - 5 930
BC036716.2 - 6 1039 4900170

View Full Product Details