Gene Transcripts

Gene Symbol
PLEK2
Entrez Gene ID
26499
Gene Name
pleckstrin 2
Gene Aliases
Chromosome Location
Chr. 14: 67386979 - 67412200 on Build GRCh38
UniGene ID
Hs.170473
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016445.2 NP_057529.1 5 705
XM_011536644.2 XP_011534946.1 5 679
GenBank mRNA AF228603.1 - 5 616
BC001226.1 - 5 649 3457876
BC008056.2 - 5 559

View Full Product Details