Gene Transcripts

Gene Symbol
MYLIP
Entrez Gene ID
29116
Gene Name
myosin regulatory light chain interacting protein
Gene Aliases
IDOL, MIR
Chromosome Location
Chr. 6: 16129086 - 16151015 on Build GRCh38
UniGene ID
Hs.484738
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013262.3 NP_037394.2 3, 4 699
XM_005249032.2 XP_005249089.1 3, 4 699
XM_005249033.2 XP_005249090.1 2, 3 423
XM_017010789.1 XP_016866278.1 3, 4 699
GenBank mRNA AF006003.1 - 3, 4 612
AF187016.2 - 3, 4 636
AF212221.1 - 2, 3 423
AF258586.1 - 3, 4 676
AK074391.1 - 3, 4 699
AK314648.1 - 3, 4 699
BC002860.2 - 3, 4 680 3638617
BT007055.1 - 3, 4 462

View Full Product Details