Gene Transcripts

Gene Symbol
LRP12
Entrez Gene ID
29967
Gene Name
LDL receptor related protein 12
Gene Aliases
ST7
Chromosome Location
Chr. 8: 104489231 - 104589024 on Build GRCh38
UniGene ID
Hs.600630
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135703.2 NP_001129175.1 6 2597
NM_013437.4 NP_038465.1 7 2654
GenBank mRNA AB208957.1 - 5 2329
AF166350.1 - 7 2569
AK126348.1 - 2 1880
AK289752.1 - 7 2558
AK299374.1 - 6 2594
BC032109.2 - 7 2530 4842797

View Full Product Details