Gene Transcripts

Gene Symbol
ADGRE2
Entrez Gene ID
30817
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
CD312, EMR2, VBU
Chromosome Location
Chr. 19: 14732697 - 14778541 on Build GRCh38
UniGene ID
Hs.531619
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271052.1 NP_001257981.1 11 1336
NM_013447.3 NP_038475.2 12 1621
XM_011527948.2 XP_011526250.1 11 1250
XM_011527949.2 XP_011526251.1 11 1250
XM_011527951.2 XP_011526253.1 10 1103
XM_011527952.2 XP_011526254.1 11 1250
XM_011527953.2 XP_011526255.1 9 1057
XM_011527954.2 XP_011526256.1 8 1788
XM_011527955.2 XP_011526257.1 6 667
XM_017026726.1 XP_016882215.1 10 1219
XM_017026727.1 XP_016882216.1 10 1103
GenBank mRNA AF114491.1 - 11 1234
AK024426.1 - 4 388
AK222891.1 - 11 1252
AK291776.1 - 11 1367
AK298690.1 - 7 696
AK298700.1 - 11 1252
AK301458.1 - 10 1103
BC034949.1 - 11 2056
BC053933.1 - 11 2049
BC082991.1 - 11 1211
BC127004.1 - 12 1617 40108332
BC127005.1 - 12 1617 40108333
CR936819.1 - 6 631

View Full Product Details