Gene Transcripts

Gene Symbol
KCNIP1
Entrez Gene ID
30820
Gene Name
potassium voltage-gated channel interacting protein 1
Gene Aliases
KCHIP1, VABP
Chromosome Location
Chr. 5: 170353055 - 170736632 on Build GRCh38
UniGene ID
Hs.484111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034837.2 NP_001030009.1 6 983
NM_001034838.2 NP_001030010.1 5 830
NM_001278339.1 NP_001265268.1 5 1025
NM_001278340.1 NP_001265269.1 6 1112
NM_014592.3 NP_055407.1 5 950
XM_017009407.1 XP_016864896.1 6 954
GenBank mRNA AF199597.1 - 5 413
AK074390.1 - 5 546
AK301775.1 - 5 605
AY170821.1 - 5 626
AY780424.1 - 6 788
BC035032.1 - 5 955
BC050375.1 - 5 717 5752207
DQ148476.1 - 5 440
DQ148477.1 - 5 413
DQ148478.1 - 6 446
DQ148479.1 - 5 488

View Full Product Details