Gene Transcripts

Gene Symbol
EHD3
Entrez Gene ID
30845
Gene Name
EH domain containing 3
Gene Aliases
PAST3
Chromosome Location
Chr. 2: 31234014 - 31269451 on Build GRCh38
UniGene ID
Hs.368808
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014600.2 NP_055415.1 2 896
GenBank mRNA AF181264.1 - 2 332
AF214736.1 - 2 573
AK294222.1 - 2 781
AK316006.1 - 2 884

View Full Product Details