Gene Transcripts

Gene Symbol
HSD17B11
Entrez Gene ID
51170
Gene Name
hydroxysteroid 17-beta dehydrogenase 11
Gene Aliases
17-BETA-HSD11, 17-BETA-HSDXI, 17BHSD11, DHRS8, PAN1B, RETSDR2, SDR16C2
Chromosome Location
Chr. 4: 87336522 - 87391303 on Build GRCh38
UniGene ID
Hs.594923
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016245.4 NP_057329.3 6 969
GenBank mRNA AF126780.1 - 6 924
AF273056.1 - 6 822
AK075348.1 - 6 853
AY358553.1 - 6 836
BC008650.1 - 6 859 3854750
BC014327.1 - 6 835 3961721
BC016367.1 - 6 866 4133318
BC021673.1 - 6 840 3827935
BC036001.1 - 6 860 4715042

View Full Product Details