Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134779.1 | NP_001128251.1 | 11 | 1306 | |
NM_016338.4 | NP_057422.3 | 11 | 1246 | ||
GenBank mRNA | AF111109.1 | - | 10 | 1056 | |
AK001696.1 | - | 11 | 1228 | ||
AK302781.1 | - | 11 | 1306 | ||
BC031694.1 | - | 3 | 307 | ||
BC033776.1 | - | 11 | 1208 | 4525238 |