Gene Transcripts

Gene Symbol
ANAPC7
Entrez Gene ID
51434
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7
Chromosome Location
Chr. 12: 110372900 - 110403730 on Build GRCh38
UniGene ID
Hs.719935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137664.1 NP_001131136.1 5 756
NM_016238.2 NP_057322.2 5 756
XM_005253903.3 XP_005253960.1 4 622
XM_011538454.2 XP_011536756.1 5 984
XM_017019425.1 XP_016874914.1 5 756
XM_017019426.1 XP_016874915.1 5 751
XM_017019427.1 XP_016874916.1 5 1550
XM_017019428.1 XP_016874917.1 4 639
GenBank mRNA AF191340.1 - 5 653
AK021809.1 - 5 735
BC009498.2 - 3 326
BC017316.2 - 1 102
BC040130.1 - 5 671
BC098264.1 - 5 652
BC098295.1 - 5 652
BC098362.1 - 5 652
BC099732.1 - 5 652
BC111798.1 - 5 652
BC141848.1 - 5 706 8860346
BC148236.2 - 5 653 40024981

View Full Product Details