Gene Transcripts
- Gene Symbol
- LARS
- Entrez Gene ID
- 51520
- Gene Name
- leucyl-tRNA synthetase
- Gene Aliases
- HSPC192, ILFS1, LARS1, LEURS, LEUS, LFIS, LRS, PIG44, RNTLS, hr025Cl
- Chromosome Location
- Chr. 5: 146113026 - 146182787 on Build GRCh38
- UniGene ID
- Hs.432674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317964.1 |
NP_001304893.1
|
10, 11 |
1318 |
|
NM_001317965.1 |
NP_001304894.1
|
10, 11 |
1337 |
|
NM_016460.3 |
NP_057544.2
|
10, 11 |
1375 |
|
NM_020117.10 |
NP_064502.9
|
11, 12 |
1456 |
|
XM_011537656.2 |
XP_011535958.1
|
11, 12 |
1322 |
|
GenBank mRNA |
AB037773.1 |
- |
11, 12 |
1329 |
|
AK023777.1 |
- |
4, 5 |
440 |
|
AK025441.1 |
- |
11, 12 |
1292 |
|
AK225372.1 |
- |
10, 11 |
1154 |
|
AK293750.1 |
- |
10, 11 |
1205 |
|
AK295874.1 |
- |
10, 11 |
1173 |
|
AK304134.1 |
- |
10, 11 |
1134 |
|
AK315178.1 |
- |
11, 12 |
1305 |
|
BC150213.1 |
- |
11, 12 |
1307 |
40148958 |
BC151214.1 |
- |
11, 12 |
1257 |
40148007 |
BC152422.1 |
- |
11, 12 |
1329 |
8862534 |
D84223.1 |
- |
11, 12 |
1234 |
|
View Full Product Details