Gene Transcripts

Gene Symbol
SF3B6
Entrez Gene ID
51639
Gene Name
splicing factor 3b subunit 6
Gene Aliases
CGI-110, HSPC175, Ht006, P14, SAP14, SAP14a, SF3B14, SF3B14a
Chromosome Location
Chr. 2: 24067584 - 24076444 on Build GRCh38
UniGene ID
Hs.177861
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016047.3 NP_057131.1 3 382
GenBank mRNA AF151868.1 - 3 378
AF161523.1 - 3 276
AF184213.1 - 3 378
AF401310.1 - 3 167
AK000621.1 - 3 282
AK311118.1 - 2 169
AK311908.1 - 3 315
BC015463.1 - 3 382 3914184
BG200650.1 - 2 143

View Full Product Details