Gene Transcripts

Gene Symbol
TRMT13
Entrez Gene ID
54482
Gene Name
tRNA methyltransferase 13 homolog
Gene Aliases
CCDC76
Chromosome Location
Chr. 1: 100133080 - 100150498 on Build GRCh38
UniGene ID
Hs.729095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019083.2 NP_061956.2 10 1186
NR_135078.1 10 945
XM_005270945.3 XP_005271002.1 10 1214
XM_005270946.2 XP_005271003.1 9 1106
XM_017001504.1 XP_016856993.1 10 2540
XM_017001505.1 XP_016856994.1 10 2529
GenBank mRNA AK002081.1 - 10 1173
AK298941.1 - 10 930
BC075811.1 - 10 1183 5104751

View Full Product Details