Gene Transcripts

Gene Symbol
TMEM39B
Entrez Gene ID
55116
Gene Name
transmembrane protein 39B
Gene Aliases
Chromosome Location
Chr. 1: 32072031 - 32102866 on Build GRCh38
UniGene ID
Hs.25544
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319677.1 NP_001306606.1 7 1372
NM_001319678.1 NP_001306607.1 5 867
NM_001319679.1 NP_001306608.1 5 864
NM_018056.3 NP_060526.2 7 1103
XM_005270988.2 XP_005271045.1 7 1225
XM_006710724.1 XP_006710787.1 7 1103
XM_006710725.3 XP_006710788.1 7 1103
XM_011541681.2 XP_011539983.1 7 1103
XM_011541683.1 XP_011539985.1 5 727
XM_011541684.2 XP_011539986.1 6 1816
XM_017001595.1 XP_016857084.1 5 849
GenBank mRNA AK001177.1 - 7 1074
AK023331.1 - 7 1100
AK091861.1 - 7 1372
AK296653.1 - 6 881
AK298766.1 - 5 775
AK300293.1 - 8 1089
AL136695.1 - 7 1081
BC004364.1 - 5 820
CR457311.1 - 6 954
CR533536.1 - 6 954

View Full Product Details