Gene Transcripts

Gene Symbol
FANCI
Entrez Gene ID
55215
Gene Name
Fanconi anemia complementation group I
Gene Aliases
KIAA1794
Chromosome Location
Chr. 15: 89243954 - 89317132 on Build GRCh38
UniGene ID
Hs.513126
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113378.1 NP_001106849.1 27, 28 3098
NM_018193.2 NP_060663.2 26, 27 2918
XM_011521756.2 XP_011520058.1 27, 28 3122
XM_011521757.2 XP_011520059.1 27, 28 3118
XM_011521764.2 XP_011520066.1 26, 27 2942
XM_011521767.2 XP_011520069.1 27, 28 3092
GenBank mRNA AB058697.1 - 13, 14 3022
AK001581.1 - 20, 21 2307
AK027564.1 - 15, 16 1805
AK055176.1 - 26, 27 2877
BC021859.1 - 4, 5 440
BC140769.1 - 27, 28 3055 9021637
BC144483.1 - 27, 28 3055 9053010
EF469766.1 - 26, 27 3008
EF567077.1 - 26, 27 3008

View Full Product Details