Gene Transcripts

Gene Symbol
SLC29A3
Entrez Gene ID
55315
Gene Name
solute carrier family 29 member 3
Gene Aliases
ENT3, HCLAP, HJCD, PHID
Chromosome Location
Chr. 10: 71319253 - 71363390 on Build GRCh38
UniGene ID
Hs.438419
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174098.1 NP_001167569.1 6 1116
NM_018344.5 NP_060814.4 6 1110
NR_033413.1 5 1027
NR_033414.1 4 800
XM_006717910.3 XP_006717973.1 6 1502
XM_017016377.1 XP_016871866.1 5 1324
XM_017016378.1 XP_016871867.1 4 1100
GenBank mRNA AF326987.1 - 5 1058
AK002022.1 - 5 861
AK304503.1 - 5 997
AK314497.1 - 6 1078
AK316152.1 - 5 1012
AX775915.1 - 6 1110
AX775917.1 - 6 1110
AX775919.1 - 6 1110
AX775921.1 - 6 1110
AY288928.1 - 6 1070
AY358686.1 - 6 1088
BC000223.1 - 3 485
BC041575.1 - 5 998
BC120996.1 - 6 1088 40116513
BC120997.1 - 6 1088 40116514

View Full Product Details