Gene Transcripts

Gene Symbol
STYK1
Entrez Gene ID
55359
Gene Name
serine/threonine/tyrosine kinase 1
Gene Aliases
NOK, SuRTK106
Chromosome Location
Chr. 12: 10618923 - 10674318 on Build GRCh38
UniGene ID
Hs.24979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018423.2 NP_060893.2 3 551
XM_005253417.2 XP_005253474.1 2 451
XM_011520736.1 XP_011519038.1 4 354
XM_011520737.2 XP_011519039.1 3 391
XM_011520738.2 XP_011519040.1 3 546
XM_017019579.1 XP_016875068.1 3 359
GenBank mRNA AF251059.1 - 3 551
AK313906.1 - 3 286
AL353940.1 - 3 299
AY563054.1 - 1 30
BC093822.1 - 1 52 7939667
BC093824.1 - 1 52 7939669
KP729000.1 - 1 30

View Full Product Details