Gene Transcripts

Gene Symbol
CTTNBP2NL
Entrez Gene ID
55917
Gene Name
CTTNBP2 N-terminal like
Gene Aliases
Chromosome Location
Chr. 1: 112391085 - 112461164 on Build GRCh38
UniGene ID
Hs.744100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018704.2 NP_061174.1 6 2100
XM_011541781.2 XP_011540083.1 6 2171
XM_017001806.1 XP_016857295.1 6 2538
GenBank mRNA AK022544.1 - 5 2021
BC016029.1 - 6 2040 3921313
DA238082.1 - 4 560

View Full Product Details