Gene Transcripts

Gene Symbol
SLC25A40
Entrez Gene ID
55972
Gene Name
solute carrier family 25 member 40
Gene Aliases
MCFP
Chromosome Location
Chr. 7: 87834499 - 87876377 on Build GRCh38
UniGene ID
Hs.208414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018843.3 NP_061331.2 8 982
GenBank mRNA AF125531.1 - 9 955
AK223020.1 - 8 936
AK290848.1 - 8 757
BC027322.1 - 7 714 4345193

View Full Product Details