Gene Transcripts

Gene Symbol
FEM1C
Entrez Gene ID
56929
Gene Name
fem-1 homolog C
Gene Aliases
EUROIMAGE686608, EUROIMAGE783647, FEM1A
Chromosome Location
Chr. 5: 115520908 - 115544919 on Build GRCh38
UniGene ID
Hs.47367
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020177.2 NP_064562.1 3 1701
XM_005272035.4 XP_005272092.1 3 1703
XM_017009647.1 XP_016865136.1 4 1787
GenBank mRNA AB058688.1 - 2 1229
AF391093.1 - 2 1139
AK025265.1 - 1 199
AK315803.1 - 3 1479
AL365409.1 - 1 241
AL831817.1 - 1 306
AY249188.1 - 2 1139

View Full Product Details