Gene Transcripts

Gene Symbol
EXOC4
Entrez Gene ID
60412
Gene Name
exocyst complex component 4
Gene Aliases
SEC8, SEC8L1, Sec8p
Chromosome Location
Chr. 7: 133253067 - 134067137 on Build GRCh38
UniGene ID
Hs.321273
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037126.1 NP_001032203.1 6 872
NM_021807.3 NP_068579.3 6 872
XM_005250523.4 XP_005250580.1 6 856
XM_017012495.1 XP_016867984.1 6 856
GenBank mRNA AB051486.1 - 6 814
AF380839.1 - 6 869
AK027688.1 - 6 866
AK295404.1 - 6 880
AL831989.2 - 6 867
AL834475.1 - 3 199
BC026174.1 - 6 848
BC067263.1 - 6 849 6183004

View Full Product Details