Gene Transcripts

Gene Symbol
ZBED8
Entrez Gene ID
63920
Gene Name
zinc finger BED-type containing 8
Gene Aliases
Buster3, C5orf54
Chromosome Location
Chr. 5: 160393148 - 160400097 on Build GRCh38
UniGene ID
Hs.529464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303251.1 NP_001290180.1 3 1421
NM_022090.4 NP_071373.2 2 1347
GenBank mRNA AK026754.1 - 1 1153
BC032603.1 - 3 1368 5535434
BC112128.1 - 1 889 8327596
BC112130.1 - 1 889 8327598

View Full Product Details