Gene Transcripts
- Gene Symbol
- ALDH8A1
- Entrez Gene ID
- 64577
- Gene Name
- aldehyde dehydrogenase 8 family member A1
- Gene Aliases
- ALDH12, DJ352A20.2
- Chromosome Location
- Chr. 6: 134917390 - 134950122 on Build GRCh38
- UniGene ID
- Hs.486520
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193480.1 |
NP_001180409.1
|
3, 4 |
521 |
|
NM_022568.3 |
NP_072090.1
|
4, 5 |
671 |
|
NM_170771.2 |
NP_739577.1
|
4, 5 |
671 |
|
GenBank mRNA |
AF303134.1 |
- |
4, 5 |
662 |
|
AK074266.1 |
- |
4, 5 |
664 |
|
AK222848.1 |
- |
4, 5 |
664 |
|
AK290784.1 |
- |
4, 5 |
618 |
|
AK298325.1 |
- |
3, 4 |
483 |
|
BC113862.1 |
- |
4, 5 |
641 |
40033493 |
BC114473.1 |
- |
5, 6 |
679 |
40033488 |
View Full Product Details