Gene Transcripts
- Gene Symbol
- ASPSCR1
- Entrez Gene ID
- 79058
- Gene Name
- ASPSCR1, UBX domain containing tether for SLC2A4
- Gene Aliases
- ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
- Chromosome Location
- Chr. 17: 81977550 - 82018929 on Build GRCh38
- UniGene ID
- Hs.298351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001251888.1 |
NP_001238817.1
|
2, 3 |
246 |
|
NM_024083.3 |
NP_076988.1
|
2, 3 |
246 |
|
XM_011523601.2 |
XP_011521903.1
|
2, 3 |
246 |
|
XM_011523602.2 |
XP_011521904.1
|
2, 3 |
246 |
|
XM_017025036.1 |
XP_016880525.1
|
2, 3 |
246 |
|
XM_017025037.1 |
XP_016880526.1
|
2, 3 |
246 |
|
XM_017025042.1 |
XP_016880531.1
|
2, 3 |
246 |
|
GenBank mRNA |
AF324219.2 |
- |
2, 3 |
171 |
|
AK057403.1 |
- |
2, 3 |
246 |
|
AK226086.1 |
- |
2, 3 |
119 |
|
AK290624.1 |
- |
2, 3 |
162 |
|
BC006152.1 |
- |
2, 3 |
201 |
3357489 |
BC018722.1 |
- |
2, 3 |
151 |
4640076 |
View Full Product Details