Gene Transcripts

Gene Symbol
ASPSCR1
Entrez Gene ID
79058
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Gene Aliases
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
Chromosome Location
Chr. 17: 81977550 - 82018929 on Build GRCh38
UniGene ID
Hs.298351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251888.1 NP_001238817.1 3, 4 369
NM_024083.3 NP_076988.1 3, 4 369
NR_045351.1 2, 3 313
XM_011523601.2 XP_011521903.1 3, 4 369
XM_011523602.2 XP_011521904.1 3, 4 369
XM_017025036.1 XP_016880525.1 3, 4 369
XM_017025037.1 XP_016880526.1 3, 4 369
XM_017025038.1 XP_016880527.1 4, 5 380
XM_017025039.1 XP_016880528.1 4, 5 297
XM_017025040.1 XP_016880529.1 3, 4 234
XM_017025041.1 XP_016880530.1 2, 3 237
XM_017025042.1 XP_016880531.1 3, 4 369
XM_017025043.1 XP_016880532.1 2, 3 237
XM_017025044.1 XP_016880533.1 4, 5 380
GenBank mRNA AF324219.2 - 3, 4 294
AK057403.1 - 3, 4 369
AK057851.1 - 2, 3 221
AK226086.1 - 3, 4 242
AK290624.1 - 3, 4 285
BC006152.1 - 3, 4 324 3357489
BC018722.1 - 3, 4 274 4640076

View Full Product Details