Gene Transcripts

Gene Symbol
OR51B4
Entrez Gene ID
79339
Gene Name
olfactory receptor family 51 subfamily B member 4
Gene Aliases
HOR5'Beta1
Chromosome Location
Chr. 11: 5301014 - 5301946 on Build GRCh38
UniGene ID
Hs.679499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033179.2 NP_149419.2 1 474
GenBank mRNA BC069094.1 - 1 474
BC100919.1 - 1 473
BC148186.1 - 1 474 40025615
BC153815.1 - 1 474 40003856

View Full Product Details