Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024551.2 | NP_078827.2 | 3 | 444 | |
XM_005253789.1 | XP_005253846.1 | 3 | 566 | ||
XM_006719018.2 | XP_006719081.1 | 4 | 563 | ||
XM_011521024.2 | XP_011519326.1 | 4 | 574 | ||
GenBank mRNA | AK127196.1 | - | 4 | 499 | |
AK128511.1 | - | 3 | 498 | ||
AY424280.1 | - | 2 | 323 | ||
BC051858.2 | - | 3 | 444 | 6527208 |