Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287437.1 | NP_001274366.1 | 6 | 764 | |
NM_001287439.1 | NP_001274368.1 | 6 | 918 | ||
NM_001287440.1 | NP_001274369.1 | 6 | 715 | ||
NM_001287441.1 | NP_001274370.1 | 6 | 679 | ||
NM_001287442.1 | NP_001274371.1 | 6 | 672 | ||
NM_001287443.1 | NP_001274372.1 | 6 | 676 | ||
NM_024900.4 | NP_079176.2 | 6 | 800 | ||
NM_199320.3 | NP_955352.1 | 6 | 800 | ||
XM_005263232.1 | XP_005263289.1 | 6 | 642 | ||
XM_017008626.1 | XP_016864115.1 | 6 | 757 | ||
XM_017008627.1 | XP_016864116.1 | 6 | 804 | ||
GenBank mRNA | AB058710.1 | - | 2 | 100 | |
AF520952.1 | - | 6 | 697 | ||
AK027620.1 | - | 6 | 720 | ||
AK074986.1 | - | 1 | 27 | ||
AK127326.1 | - | 6 | 572 | ||
AK304235.1 | - | 6 | 633 | ||
AK304815.1 | - | 6 | 589 | ||
BC032376.1 | - | 6 | 634 | 5111727 |