Gene Transcripts

Gene Symbol
FOXRED2
Entrez Gene ID
80020
Gene Name
FAD dependent oxidoreductase domain containing 2
Gene Aliases
ERFAD
Chromosome Location
Chr. 22: 36487186 - 36507101 on Build GRCh38
UniGene ID
Hs.387601
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102371.1 NP_001095841.1 7 1659
NM_024955.5 NP_079231.4 7 1698
XM_017028941.1 XP_016884430.1 6 1331
XM_017028942.1 XP_016884431.1 6 1428
GenBank mRNA AK025714.1 - 7 1598
AK026975.1 - 4 779
AK091053.1 - 2 590
AK225347.1 - 7 1598
AK315555.1 - 7 1601
BC015726.1 - 7 1582 4866761
BC027716.2 - 7 1677 4127473
BC040351.1 - 7 1621 3873448
CR456444.1 - 6 1673

View Full Product Details