Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193478.1 | NP_001180407.1 | 7 | 1332 | |
NM_030911.3 | NP_112173.1 | 7 | 1332 | ||
NR_036439.1 | 6 | 1154 | |||
XM_006719871.3 | XP_006719934.1 | 6 | 1101 | ||
XM_011535250.2 | XP_011533552.1 | 7 | 1296 | ||
XM_011535251.1 | XP_011533553.1 | 7 | 1276 | ||
XM_011535253.1 | XP_011533555.1 | 7 | 1228 | ||
XM_017020778.1 | XP_016876267.1 | 6 | 1035 | ||
XM_017020779.1 | XP_016876268.1 | 8 | 1452 | ||
XM_017020780.1 | XP_016876269.1 | 8 | 1799 | ||
XM_017020781.1 | XP_016876270.1 | 7 | 1271 | ||
XM_017020782.1 | XP_016876271.1 | 8 | 1375 | ||
XM_017020783.1 | XP_016876272.1 | 6 | 1496 | ||
XM_017020784.1 | XP_016876273.1 | 5 | 1067 | ||
GenBank mRNA | AK297400.1 | - | 6 | 1118 | |
AK312835.1 | - | 7 | 1302 | ||
AY027525.1 | - | 7 | 1281 | ||
BC048092.1 | - | 8 | 1300 | ||
BC125202.1 | - | 7 | 1246 | 40123430 |