Gene Transcripts

Gene Symbol
CCDC115
Entrez Gene ID
84317
Gene Name
coiled-coil domain containing 115
Gene Aliases
CDG2O, ccp1
Chromosome Location
Chr. 2: 130337933 - 130342681 on Build GRCh38
UniGene ID
Hs.104203
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321118.1 NP_001308047.1 3 689
NM_001321119.1 NP_001308048.1 3 211
NM_032357.3 NP_115733.2 3 510
NR_104471.1 3 689
NR_135548.1 3 510
GenBank mRNA AK054693.1 - 3 476
AK124405.1 - 1 423
AK295922.1 - 3 346
BC006429.1 - 3 272 3350642
BG179982.1 - 211
DB025106.1 - 3 511

View Full Product Details