Gene Transcripts

Gene Symbol
RAB11FIP4
Entrez Gene ID
84440
Gene Name
RAB11 family interacting protein 4
Gene Aliases
RAB11-FIP4
Chromosome Location
Chr. 17: 31391624 - 31538217 on Build GRCh38
UniGene ID
Hs.406788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303542.2 NP_001290471.2 10 1376
NM_032932.5 NP_116321.2 12 1669
GenBank mRNA AB058724.1 - 12 1358
AJ314646.2 - 12 1358
AK074341.1 - 5 447
AK097424.1 - 10 1319
AL831830.1 - 4 418
AY169244.1 - 12 1669
BC093914.1 - 12 1472 7939759
BC101517.1 - 12 1472 8069023

View Full Product Details