Gene Transcripts
- Gene Symbol
- ZNF512
- Entrez Gene ID
- 84450
- Gene Name
- zinc finger protein 512
- Gene Aliases
- Chromosome Location
- Chr. 2: 27582969 - 27623215 on Build GRCh38
- UniGene ID
- Hs.529178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271286.1 |
NP_001258215.1
|
9, 10 |
1177 |
|
NM_001271287.1 |
NP_001258216.1
|
9, 10 |
1205 |
|
NM_001271288.1 |
NP_001258217.1
|
9, 10 |
1202 |
|
NM_001271289.1 |
NP_001258218.1
|
9, 10 |
1166 |
|
NM_001271318.1 |
NP_001258247.1
|
11, 12 |
1365 |
|
NM_032434.3 |
NP_115810.2
|
10, 11 |
1264 |
|
GenBank mRNA |
AB058708.1 |
- |
8, 9 |
1023 |
|
AK057028.1 |
- |
10, 11 |
1174 |
|
AK293771.1 |
- |
9, 10 |
1080 |
|
AK294055.1 |
- |
10, 11 |
1370 |
|
AK299818.1 |
- |
9, 10 |
1067 |
|
AK316011.1 |
- |
11, 12 |
1225 |
|
AK316299.1 |
- |
9, 10 |
1119 |
|
AL833748.1 |
- |
2, 3 |
246 |
|
BC043221.2 |
- |
9, 10 |
1113 |
5295402 |
BC093041.1 |
- |
9, 10 |
1132 |
6173398 |
BX647996.1 |
- |
9, 10 |
1111 |
|
View Full Product Details