Gene Transcripts

Gene Symbol
CARD6
Entrez Gene ID
84674
Gene Name
caspase recruitment domain family member 6
Gene Aliases
CINCIN1
Chromosome Location
Chr. 5: 40841190 - 40855354 on Build GRCh38
UniGene ID
Hs.200242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032587.3 NP_115976.2 3 2045
XM_017009989.1 XP_016865478.1 2 1598
XM_017009990.1 XP_016865479.1 4 2279
GenBank mRNA AB209844.1 - 3 1865
AF356193.1 - 3 2169
BC093825.1 - 3 1992 7939670

View Full Product Details