Gene Transcripts

Gene Symbol
FAM71F1
Entrez Gene ID
84691
Gene Name
family with sequence similarity 71 member F1
Gene Aliases
FAM137A, NYD-SP18
Chromosome Location
Chr. 7: 128709061 - 128731743 on Build GRCh38
UniGene ID
Hs.732440
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282788.1 NP_001269717.1 6 966
NM_001282789.1 NP_001269718.1 7 913
NM_032599.3 NP_115988.1 6 972
NR_104242.1 7 1270
NR_104243.1 8 1159
GenBank mRNA AF367470.1 - 6 925
AK093161.1 - 7 910
AK313608.1 - 6 917
BC037248.1 - 6 905 5165912

View Full Product Details