Gene Transcripts

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
Chromosome Location
Chr. 10: 87751512 - 87841345 on Build GRCh38
UniGene ID
Hs.435948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321967.1 NP_001308896.1 7 806
NM_001321969.1 NP_001308898.1 8 896
NM_032810.3 NP_116199.2 7 1178
NR_135914.1 7 806
XM_005270252.4 XP_005270309.1 7 900
XM_011540302.1 XP_011538604.1 5 571
XM_011540303.2 XP_011538605.1 7 853
XM_017016847.1 XP_016872336.1 7 1047
XM_017016849.1 XP_016872338.1 8 1019
XM_017016850.1 XP_016872339.1 8 1070
GenBank mRNA AF361493.1 - 6 1012
AK027506.1 - 4 406
AK075223.1 - 7 1107
AL834370.1 - 6 591
BC010868.1 - 3 241
BC063530.1 - 6 637
BC073998.1 - 6 749 6199813

View Full Product Details